Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleate, 0.2M buffer soln., pH 6.5, Thermo Scientific™

Catalog No. AAJ61156AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61156AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61156AK Supplier Thermo Scientific Chemicals Supplier No. J61156AK

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-46-0
Molecular Formula C4H4Na2O5
Molecular Weight (g/mol) 178.051
MDL Number MFCD00012466
InChI Key WPUMTJGUQUYPIV-UHFFFAOYSA-L
PubChem CID 8736
IUPAC Name disodium;2-hydroxybutanedioate
SMILES C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]

Specifications

Concentration 0.2 M
pH 6.5
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material Maleate buffer soln.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.