Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maleic Acid, NF, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18604147 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum

M1203, 110-16-7, C4H4O4

Spectrum™ Chemical Maleic Acid, NF

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

CAS Max % 100%
UN Number UN3261
Percent Purity 99 to 101%
Grade NF
Quantity 100 g
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.