Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Maleic anhydride

Catalog No. 503125803 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5803 1 kg
50-312-6140 25 kg
50-312-5771 5 kg
50-312-5804 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-312-5803 Supplier Sigma Aldrich Supplier No. M1881KGA
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Maleic anhydride

Specifications

CAS 108-31-6
Melting Point 51°C to 56°C
Boiling Point 200°C
Molecular Formula C4H2O3
Linear Formula C4H2O3
Quantity 1 kg
MDL Number MFCD00005518
Synonym 2,5-Furandione
Molecular Weight (g/mol) 98.06
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.