Learn More
Maleic anhydride, 98+%
Description
Maleic anhydride is used in the preparation of unsaturated polyester resins, 4-butanediol, fumairc acid, tetrahydrofuran and gamma-butyrolactone. It is also used as lubricating oil additives, dispersants, artificial sweeteners, flavor enhancements, floor polishes and corrosion inhibitors. It finds application in personal care products like hair sprays. Further, it is used in water treatment chemicals, detergents, pharmaceuticals and copolymers. It is an active substrate for Diels-Alder reactions and dimerizes to form cyclobutane tetracarboxylic dianhydride .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Maleic anhydride is used in the preparation of unsaturated polyester resins, 4-butanediol, fumairc acid, tetrahydrofuran and gamma-butyrolactone. It is also used as lubricating oil additives, dispersants, artificial sweeteners, flavor enhancements, floor polishes and corrosion inhibitors. It finds application in personal care products like hair sprays. Further, it is used in water treatment chemicals, detergents, pharmaceuticals and copolymers. It is an active substrate for Diels-Alder reactions and dimerizes to form cyclobutane tetracarboxylic dianhydride .
Solubility
Soluble in water, acetone and dioxane. Slightly soluble in methanol, chloroform and benzene.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, strong reducing agents, alkali metals and amines.
Specifications
Specifications
| Melting Point | 52°C to 56°C |
| Boiling Point | 202°C |
| Flash Point | 103°C (217°F) |
| Assay Percent Range | ≥98% |
| Quantity | 250 g |
| UN Number | UN2215 |
| Beilstein | 106909 |
| Sensitivity | Moisture sensitive |
| Merck Index | 14,5704 |
| Solubility Information | Soluble in water,acetone and dioxane. Slightly soluble in methanol,chloroform and benzene. |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H317-H334-H372-H500 |
| Hazard Statement | GHS H Statement H334-H314-H302-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: MALEIC ANHYDRIDE |
| EINECSNumber | 203-571-6 |
| RTECSNumber | ON3675000 |
| TSCA | Yes |
| storageNote1 | Moisture Sensitive |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.