Learn More
Thermo Scientific™ Imject™ Maleimide-Activated Blue Carrier™ Protein
Catalog No. pi77661
Click to view available options
Quantity:
2 mg
10 mg
10 x 10 mg
Make hapten-carrier immunogens comprising cysteine-containing peptides and other thiolated antigens with sulfhydryl-reactive immunogenic hemocyanin (CCH).
Thermo Scientific™ Maleimide-Activated Blue Carrier Protein is sulfhydryl-reactive immunogenic hemocyanin (CCH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Blue Carrier Protein is a large homocyanin that is highly immunogenic like keyhole limpet hemocyanin (KLH) yet is more soluble. The carrier is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Numerous antigen molecules can be conjugated to each Blue Carrier Protein molecule to make them highly immunogenic and increase the likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest.
Highlights:
  • Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of carrier, ensuring consistently high levels of conjugatio
  • Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
  • Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
  • High-quality carrier protein – Blue Carrier Protein is ultrapure Concholepas concholepas hemocyanin (CCH) prepared from molluscan hemolymph
  • Highly immunogenic and soluble – CCH has a high molecular mass that elicits a stronger immune response than BSA or ovalbumin
TRUSTED_SUSTAINABILITY
Specifications
Product Type Imject Maleimide-Activated Blue Carrier Protein
Product Line Blue Carrier™, Imject™
Content And Storage Upon receipt store at -20°C. Product is shipped on ice.
Quantity 2 mg
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.