Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malic Acid Standard, 10,000 ppm C4H6O5 in 10% (v/v) Ethanol, Ricca Chemical
SDP

Catalog No. r4584100100
Click to view available options
Quantity:
100 mL

Chemical Identifiers

CAS 97-67-6, 64-17-5, 7732-18-5
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
Synonym l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2S)-2-hydroxybutanedioic acid
SMILES O[C@@H](CC(O)=O)C(O)=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 0.9, 7.1, 90.0
CAS Max % 0.94, 7.39, 94.0
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Malic Acid Standard, 10000ppm in 10% Ethanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.