Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Malondialdehyde Antibody (6H6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259366MFV450
Description
Malondialdehyde Monoclonal antibody specifically detects Malondialdehyde in Non-species specific samples. It is validated for Flow CytometrySpecifications
Malondialdehyde | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Non-species specific | |
Purified |
Flow Cytometry | |
6H6 | |
Flow Cytometry | |
3-Propanedial;Malonaldehyde;Malondialdehyde;MDA | |
Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction