Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Malondialdehyde Monoclonal Antibody (11E3), APC
GREENER_CHOICE

Catalog No. PIMA545805
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545805 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545805 Supplier Invitrogen™ Supplier No. MA545805
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45805 was sufficient for detection of Malondialdehyde in 2 μg of Malondialdehyde conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins.

Malondialdehyde is an organic compound with the formula CH2(CHO)2, and a byproduct of lipid metabolism in the body. Malondialdehyde, a highly reactive compound, is one of the many reactive electrophile species that cause toxic stress in cells and form covalent protein adducts, called advanced lipoxidation end products (ALE). This compound also forms mutagenic DNA adducts when it reacts with deoxyadenosine and deoxyguanosine in DNA. Malondialdehyde is also found in many foods and is present in high levels in rancid foods.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Malondialdehyde
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 11E3
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene Alias 3-Propanedial; Malonaldehyde; Malondialdehyde; MDA
Host Species Mouse
Immunogen Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.