Learn More
Invitrogen™ Malondialdehyde Monoclonal Antibody (6H6)

Description
A 1:1,000 dilution of MA5-27559 was sufficient for detection of Malondialdehyde in 2 μg of Malondialdehyde conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for Malondialdehyde conjugated proteins. Does not detect free Malondialdehyde. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Methylglyoxal modified proteins.
Specifications
Specifications
| Antigen | Malondialdehyde |
| Applications | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6H6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene Alias | 3-Propanedial; Malonaldehyde; Malondialdehyde; MDA |
| Host Species | Mouse |
| Immunogen | Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.