Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Malonic Acid
SCI_ED

Catalog No. p-4416583 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Solid

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 141-82-2
Molecular Formula C3H4O4
Molecular Weight (g/mol) 104.061
InChI Key OFOBLEOULBTSOW-UHFFFAOYSA-N
Synonym malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
PubChem CID 867
ChEBI CHEBI:30794
IUPAC Name propanedioic acid
SMILES C(C(=O)O)C(=O)O

Specifications

Color White
Density 1.63g/mL
Assay Percent Range >99%
Packaging Solid
Linear Formula HO2CCH2CO2H
Quantity 25 g
Formula Weight 104.06
Chemical Name or Material Malonic Acid
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.