Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MALRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565158 Supplier Invitrogen™ Supplier No. PA565158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSGSKEGSVAR ITTSKSFPAS LGMCTVRFWF YMIDPRSMGI LKVYTIEESG LNILVWSVIG NK Highest antigen sequence identity to the following orthologs - mouse 70%, rat 33%.

Enhances production and/or transport of FGF19 and thus has a role in regulation of bile acid synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MALRD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MALRD1
Gene Accession No. Q5VYJ5
Gene Alias bA265G8.2; C10orf112; DIET1; MALRD1; MAM and LDL receptor class A domain containing 1; MAM and LDL-receptor class A domain-containing protein 1; MAM and LDL-receptor class A domain-containing protein C10orf112; novel MAM domain containing protein
Gene Symbols MALRD1
Host Species Rabbit
Immunogen Recombinant Human MALRD1. Recombinant protein control fragment (Product #RP-105907).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.