Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MALRD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565203
Description
Immunogen sequence: DSTVIWRVLYN QGKQWLEATI QLGRLSQPFH LSLDKVSLGI YDGVSAIDDI RFENCTLPLP AESCEGLDHF WCRHTRACIE KLRLCD Highest antigen sequence identity to the following orthologs - mouse 70%, rat 71%.
Enhances production and/or transport of FGF19 and thus has a role in regulation of bile acid synthesis.
Specifications
MALRD1 | |
Polyclonal | |
Unconjugated | |
MALRD1 | |
bA265G8.2; C10orf112; DIET1; MALRD1; MAM and LDL receptor class A domain containing 1; MAM and LDL-receptor class A domain-containing protein 1; MAM and LDL-receptor class A domain-containing protein C10orf112; novel MAM domain containing protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
340895 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VYJ5 | |
MALRD1 | |
Recombinant Human MALRD1. Recombinant protein control fragment (Product #RP-97226). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction