Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MALSU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554222 Supplier Invitrogen™ Supplier No. PA554222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFYVVKMYKH LKCKRDPHVK IEGKDTDDWL CVDFGSMVIH LMLPETREIY ELEKLWTLRS YDDQLAQIAP ETVPEDFILG IEDDTSSVTP V Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

Required for normal mitochondrial ribosome function and mitochondrial translation (PubMed:22238375, PubMed:23171548). May play a role in ribosome biogenesis by preventing premature association of the 28S and 39S ribosomal subunits (Probable). Interacts with mitochondrial ribosomal protein L14 (MRPL14), probably blocking formation of intersubunit bridge B8, preventing association of the 28S and 39S ribosomal subunits (Probable). Addition to isolated mitochondrial ribosomal subunits partially inhibits translation, probably by interfering with the association of the 28S and 39S ribosomal subunits and the formation of functional ribosomes (Probable). May also participate in the assembly and/or regulation of the stability of the large subunit of the mitochondrial ribosome (PubMed:22238376, PubMed:23171548). May function as a ribosomal silencing factor (Probable). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MALSU1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MALSU1
Gene Accession No. Q96EH3
Gene Alias 2410003K15Rik; C7orf30; Malsu1; Matsu1; mitochondrial assembly of ribosomal large subunit 1; mitochondrial assembly of ribosomal large subunit protein 1; mtRsfA; protein C7orf30 homolog; RGD1306936
Gene Symbols MALSU1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MALSU1. Recombinant protein control fragment (Product #RP-92767).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115416
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.