Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MALT1 Antibody (rMT1/410), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308824FR
Description
MALT1 Monoclonal antibody specifically detects MALT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MALT1 | |
Monoclonal | |
DyLight 680 | |
caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase | |
Recombinant fragment (around aa701-808) of human MALT1 protein (exact sequence is proprietary) (Uniprot: Q9UDY8) | |
100 μg | |
Apoptosis, Cancer, Tumor Suppressors | |
10892 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rMT1/410 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction