Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltol, FCC, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604170 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum

Catalog No. 18-604-170

Supplier: Spectrum Chemical Manufacturing Corporation M1189500GM

Only null left
Add to Cart
Add to Cart

M1189, 118-71-8, C6H6O3

Spectrum™ Chemical Maltol, FCC is used as a flavor enhancer in breads and cakes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 118-71-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006578
InChI Key XPCTZQVDEJYUGT-UHFFFAOYSA-N
IUPAC Name 3-hydroxy-2-methyl-4H-pyran-4-one
SMILES CC1=C(O)C(=O)C=CO1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Melting Range 160-164 C
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 99%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.