Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maltose Binding Protein Antibody (R29.6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $427.50
Specifications
Antigen | Maltose Binding Protein |
---|---|
Clone | R29.6 |
Concentration | 1 mg/mL |
Dilution | Western Blot : 1:100 - 1:2000, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Classification | Monoclonal |
Description
Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Maltose Binding Protein | |
1 mg/mL | |
Monoclonal | |
Mouse | |
PBS with 0.02% sodium azide | |
MOS maltose binding protein fusion protein | |
Primary | |
This antibody is useful for detection and isolation of recombinant MBP fusion proteins. |
R29.6 | |
Western Blot : 1:100 - 1:2000, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Unconjugated | |
Non-species specific | |
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
IgG1 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title