Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maltose Binding Protein Antibody (R29), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066609H
Description
Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Bacteria samples. It is validated for Western Blot.Specifications
Maltose Binding Protein | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
malE | |
Recombinant maltose binding protein | |
0.125 mL | |
Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
948538 | |
Bacteria | |
Purified |
Western Blot | |
R29 | |
Western Blot | |
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes maltose binding protein (MBP). The is useful in recognizing proteins expressed with an MBP tag. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction