Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maltose Binding Protein Antibody (R3.2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP276652IR
Description
Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Maltose Binding Protein | |
Monoclonal | |
DyLight 755 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
This antibody is useful for detection and isolation of recombinant MBP fusion proteins. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
R3.2 | |
Western Blot, Immunohistochemistry | |
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
MOS maltose binding protein fusion protein | |
Primary | |
Non-species specific | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction