Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maltose Binding Protein Antibody (TVG MBP 3), Novus Biologicals™

Mouse Monoclonal Antibody
$173.10 - $402.00
Specifications
Antigen | Maltose Binding Protein |
---|---|
Clone | TVG MBP 3 |
Dilution | Western Blot : 1:100 - 1:2000, ELISA, Immunoprecipitation : 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
Maltose Binding Protein | |
Western Blot : 1:100 - 1:2000, ELISA, Immunoprecipitation : 1:10 - 1:500 | |
Monoclonal | |
Purified | |
Non-species specific | |
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
IgM | |
Thiophilic Chromatography | |
This antibody is useful for detection and isolation of recombinant MBP fusion proteins. |
TVG MBP 3 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Mouse | |
PBS with 0.02% sodium azide | |
A single intravenous injection of the maltose binding protein MBP-E2. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title