Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Binding Protein/MBP Rabbit anti-Bacteria∣Bovine, Polyclonal, Bioss
SDP

Catalog No. 501985050
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5050 Supplier Bioss Supplier No. BS2967R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by the classic MBP gene is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. However, MBP-related transcripts are also present in the bone marrow and the immune system. These mRNAs arise from the long MBP gene (otherwise called 'Golli-MBP') that contains 3 additional exons located upstream of the classic MBP exons. Alternative splicing from the Golli and the MBP transcription start sites gives rise to 2 sets of MBP-related transcripts and gene products. The Golli mRNAs contain 3 exons unique to Golli-MBP, spliced in-frame to 1 or more MBP exons. They encode hybrid proteins that have N-terminal Golli aa sequence linked to MBP aa sequence. The second family of transcripts contain only MBP exons and produce the well characterized myelin basic proteins. This complex gene structure is conserved among species suggesting that the MBP transcription unit is an integral part of the Golli transcription unit and that this arrangement is important for the function and/or regulation of these genes.

Specifications

Antigen Maltose Binding Protein/MBP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MBP
Gene Accession No. P02687
Gene Alias 20 kDa microtubule-stabilizing protein; BOS_22097; C76307; golli mbp; golli-mbp; Golli-Mbp; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; LOC100344447; Mbp; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mld; Myelin A1 protein; Myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; Myelin membrane encephalitogenic protein; R75289; Shi; shiverer
Gene Symbols MBP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from E. coli strain 12 Maltose Binding Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 618684
Target Species Bacteria, Bovine
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.