Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maltose Binding Protein Rabbit anti-E. coli, Clone: S02-6F4, Novus Biologicals™

Rabbit Monoclonal Antibody
$274.50 - $586.50
Specifications
Antigen | Maltose Binding Protein |
---|---|
Clone | S02-6F4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in E. coli samples. It is validated for Western BlotSpecifications
Maltose Binding Protein | |
Western Blot | |
Unconjugated | |
Rabbit | |
Epitope Tags, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
A synthesized peptide derived from human Maltose Binding Protein (Uniprot#: P0AEX9) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S02-6F4 | |
Monoclonal | |
Purified | |
RUO | |
E. coli | |
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title