Learn More
Maltose Phosphorylase Polyclonal Antibody, Rockland™

Goat Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Maltose Phosphorylase |
---|---|
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0307
![]() ![]() |
Rockland Immunochemicals
2001012300100 |
100 μg |
Each for $636.24
|
|
|||||
50-199-0308
![]() ![]() |
Rockland Immunochemicals
200101230S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-MALTOSE PHOSPHORYLASE is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum as well as purified and partially purified Maltose Phosphorylase [E.coli]. Cross reactivity against Maltose Phosphorylase from other sources is unknown.
Maltose Phosphorylase catalyzes the phosphorolysis of maltose, leading to the formation of glucose and glucose 1-P.Specifications
Maltose Phosphorylase | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Primary | |
Antibody |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Goat | |
Bacteria | |
Maltose Phosphorylase [E.coli] | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.