Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Malvidin-3-galactoside chloride, ≥95% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mg
Description
Food and Beverages

Specifications
Specifications
CAS | 30113-37-2 |
Linear Formula | C23H25 ClO12 |
MDL Number | MFCD00185577 |
Synonym | Arthanitin chloride; Primulin |
Type | Analytical Standard |
Quantity | 1 mg |
Formula Weight | 528.89 |
Percent Purity | ≥95% (HPLC) |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction