Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAMDC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566300 Supplier Invitrogen™ Supplier No. PA566300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VCNFVCDCRDC SDEAQCGYHG ASPTLGAPFA CDFEQDPCGW RDISTSGYSW LRDRAGAALE GPGPHSDHTL GTDL Highest antigen sequence identity to the following orthologs - mouse 77%, rat 79%.

Probably involved in the sorting and selective transport of receptors and ligands across polarized epithelia. Target protein is a glycoprotein localized to the apical endosomal tubules of intestinal epithelial cells; may play a role in intestinal transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAMDC4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAMDC4
Gene Accession No. Q6UXC1
Gene Alias AEGP; apical early endosomal glycoprotein; apical endosomal glycoprotein; endotubin; MAM domain containing 4; MAM domain-containing protein 4; MAMDC4; UNQ3001/PRO9742
Gene Symbols MAMDC4
Host Species Rabbit
Immunogen Recombinant Human MAMDC4. Recombinant protein control fragment (Product #RP-106980).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.