Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAML1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519796
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519796 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519796 Supplier Invitrogen™ Supplier No. PA519796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and horse based on sequence homology.

This protein is the human homolog of mastermind, a Drosophila protein that plays a role in the Notch signaling pathway involved in cell-fate determination. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAML1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene MAML1
Gene Accession No. Q92585
Gene Alias AI644666; AW743257; D930008C07Rik; FLJ53540; KIAA0200; Mam1; Mam-1; MAML1; mastermind homolog; mastermind like 1 (Drosophila); mastermind like transcriptional coactivator 1; mastermind-like 1; mastermind-like protein 1; mastermind-like transcriptional coactivator 1; mKIAA0200
Gene Symbols MAML1
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 950 to the C-terminus MAML1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303101, 9794
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.