Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAML1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111083
Description
Immunogen sequence: QLGSPQVRAG SAGQTFLGPS SAPVSTDSPS LGGSQTLFHT SGQPRADNPS PNLMPASAQA QNAQRALAGV VLPSQGPGGA SELSSAHQLQ QIAAKQKREQ MLQNPQ.
This protein is the human homolog of mastermind, a Drosophila protein that plays a role in the Notch signaling pathway involved in cell-fate determination. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway.
Specifications
| MAML1 | |
| Polyclonal | |
| Unconjugated | |
| MAML1 | |
| AI644666; AW743257; D930008C07Rik; FLJ53540; KIAA0200; Mam1; Mam-1; MAML1; mastermind homolog; mastermind like 1 (Drosophila); mastermind like transcriptional coactivator 1; mastermind-like 1; mastermind-like protein 1; mastermind-like transcriptional coactivator 1; mKIAA0200 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9794 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92585 | |
| MAML1 | |
| Recombinant Protein corresonding to Human MAML1. Recombinant protein control fragment (Product #RP-96915). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction