Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAML1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982205
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2205 Supplier Bioss Supplier No. BS12396R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This protein is the human homolog of mastermind, a Drosophila protein that plays a role in the Notch signaling pathway involved in cell-fate determination. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway.

Specifications

Antigen MAML1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAML1
Gene Accession No. Q92585
Gene Alias AI644666; AW743257; D930008C07Rik; FLJ53540; Kiaa0200; Mam1; Mam-1; Maml1; mastermind homolog; mastermind like 1 (Drosophila); mastermind like transcriptional coactivator 1; mastermind-like 1; Mastermind-like protein 1; mastermind-like transcriptional coactivator 1; mKIAA0200
Gene Symbols MAML1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAML1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9794
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.