Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAML2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578602 Supplier Invitrogen™ Supplier No. PA578602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HeLa, HeLa nuclear extract Predicted Reactivity: Mouse (88%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1. Potentiates activation by NOTCH3 and NOTCH4 more efficiently than MAML1 or MAML3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAML2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene MAML2
Gene Accession No. Q8IZL2
Gene Alias 5930431H10; BC032967; KIAA1819; MAM2; Mam-2; MAM3; MAM-3; MAML2; mastermind like 2 (Drosophila); mastermind like transcriptional coactivator 2; mastermind-like 2; mastermind-like protein 2; mastermind-like transcriptional coactivator 2; MLL-MAML2
Gene Symbols MAML2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MAML2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.