Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAML2 Polyclonal Antibody
Supplier: Thermo Scientific PA5115538
Description

Specifications
MAML2 | |
Polyclonal | |
Unconjugated | |
MAML2 | |
5930431H10; BC032967; KIAA1819; MAM2; Mam-2; MAM3; MAM-3; MAML2; mastermind like 2 (Drosophila); mastermind like transcriptional coactivator 2; mastermind-like 2; mastermind-like protein 2; mastermind-like transcriptional coactivator 2; MLL-MAML2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
689844, 84441 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q8IZL2 | |
MAML2 | |
A synthesized peptide derived from human MAML2(Accession Q8IZL2), corresponding to amino acid residues Q1092-W1142. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction