Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAML2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116738
Description
Positive Samples: A-549, 293T, Mouse brain, Mouse lung Immunogen sequence: NQQLMGKKQT LQRQIMEQKQ QLLLQQQMLA DAEKIAPQDQ INRHLSRPPP DYKDQRRNVG NMQPTAQYSG GSSTISLNSN QALANPVSTH TILTPNSSLL S.
Acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1. Potentiates activation by NOTCH3 and NOTCH4 more efficiently than MAML1 or MAML3.
Specifications
MAML2 | |
Polyclonal | |
Unconjugated | |
MAML2 | |
5930431H10; BC032967; KIAA1819; MAM2; Mam-2; MAM3; MAM-3; MAML2; mastermind like 2 (Drosophila); mastermind like transcriptional coactivator 2; mastermind-like 2; mastermind-like protein 2; mastermind-like transcriptional coactivator 2; MLL-MAML2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
270118, 689844, 84441 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.18 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8IZL2 | |
MAML2 | |
A synthetic peptide corresponding to a sequence within amino acids 750-850 of human MAML2 (NP_1158031). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction