Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAML3 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002128UV
Description
MAML3 Polyclonal antibody specifically detects MAML3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MAML3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CAG/CTG 3, ERDA3, MAM-2, mam-3, mastermind-like 3 (Drosophila), mastermind-like protein 3, polyglutamine rich, trinucleotide repeat containing 3 | |
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human Mastermind-Like 3 using the numbering given in entry NP_061187.2 (GeneID 55534). | |
0.1 mL | |
Neuroscience | |
55534 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction