Learn More
MAML3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300684AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
MAML3 acts as a transcriptional coactivator for NOTCH proteins. The protein has been shown to amplify NOTCH-induced transcription of HES1.Specifications
| MAML3 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q96JK9 | |
| MAML3 | |
| Between 1100 and C-term | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| MAML3 | |
| Mastermind-like protein 3; Mam-3; KIAA1816; MAML3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 433586, 55534 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.