Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAML3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982206
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2206 Supplier Bioss Supplier No. BS12398R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAML3 acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1.

Specifications

Antigen MAML3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAML3
Gene Accession No. Q96JK9
Gene Alias AV234550; BC049812; CAG repeat containing (glia-derived nexin I alpha); CAG repeat domain; CAGH3; ERDA3; expanded repeat domain, CAG/CTG 3; GDN; Glrp1; glutamine repeat protein 1; KIAA1816; KIAA1816 protein-like; LOC100354708; MAM2; Mam-2; Mam-3; MAML3; mastermind like 3 (Drosophila); mastermind like transcriptional coactivator 3; mastermind-2; mastermind-like 3; Mastermind-like protein 3; mastermind-like transcriptional coactivator 3; mKIAA1816; polyglutamine rich; TNRC3; trinucleotide repeat containing 3
Gene Symbols MAML3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAML3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55534
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.