Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAMLD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118044
This item is not returnable.
View return policy
Description
MAMLD1 Polyclonal Antibody for Western Blot, IHC (P)
CXorf6 is a protein predicted based on an ORF found in chromosome 14.
Specifications
MAMLD1 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q13495 | |
MAMLD1 | |
E. coli-derived Human MAMLD1 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
MAMLD1 | |
CG1; CXorf6; F18; G630014P10Rik; HYSP2; MAMLD1; mastermind like domain containing 1; mastermind-like domain containing 1; mastermind-like domain-containing protein 1; Protein CG1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10046 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction