Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAMLD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540847 Supplier Invitrogen™ Supplier No. PA540847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LEELTKIQDP SPNELDLEKI LGTKPEEPLV LDHPQATLST TPKPSVQMSH Sequence homology: Dog: 86%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 86%; Rabbit: 86%; Rat: 79%.

CXorf6 is a protein predicted based on an ORF found in chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAMLD1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAMLD1
Gene Accession No. Q13495
Gene Alias CG1; CXorf6; F18; G630014P10Rik; HYSP2; MAMLD1; mastermind like domain containing 1; mastermind-like domain containing 1; mastermind-like domain-containing protein 1; Protein CG1
Gene Symbols MAMLD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CXORF6 (aa 180-229).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10046
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.