Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mammaglobin A Antibody (MGB/4057R), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308526AF647
Description
Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
Mammaglobin A | |
Monoclonal | |
Alexa Fluor 647 | |
mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
Recombinant full-length human Mammaglobin A protein (Uniprot: Q13296) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Protein Array | |
MGB/4057R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
4250 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction