Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mammaglobin A Monoclonal Antibody (1G8D6,2E7G9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515479
Description
MA5-15479 targets Mammaglobin-1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15479 immunogen is purified recombinant fragment of Mammaglobin-1 expressed in E. Coli..
Mammaglobin is a gene that is expressed almost exclusively in the normal breast epithelium and human breast cancer. It is a member of the secretoglobin gene family and forms a heterodimer with lipophilin B. It has been suggested that mammaglobin may be a useful marker for breast cancer clinical research. Studies investigating the detection of mRNA by RT PCR from circulating carcinoma cells in the peripheral blood of breast cancer patients have shown that mammaglobin is a highly specific marker and correlates with several prognostic factors, such as lymph node involvement.
Specifications
Mammaglobin A | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q13296 | |
SCGB2A2 | |
Purified recombinant fragment of Mammaglobin-1 expressed in E. Coli. | |
RUO | |
4250 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1G8D6,2E7G9, 2E7G9 | |
Unconjugated | |
SCGB2A2 | |
C3.1; HGNC:7050; mammaglobin 1; mammaglobin A; Mammaglobin-1; Mammaglobin-A; MGB1; MGC71974; prostatein; prostatein C3 subunit; prostatein peptide C3; prostatic steroid binding protein, allele C3(1); prostatic steroid-binding protein C3; Psbpc3; SCGB2A2; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 2; UGB2 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction