Learn More
Mammaglobin A Mouse anti-Human, Clone: 304-1A5, Cocktail, Zeta
Mouse Monoclonal Antibody Cocktail
Supplier: ZETA CORPORATION Z2007MP

Description
This product is diluted and in a ready-to-use formulation. A recommended positive control tissue for this product is Breast carcinoma, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Mammaglobin A | |
Monoclonal | |
Unconjugated | |
SCGB2A2 | |
C3.1; HGNC:7050; mammaglobin 1; mammaglobin A; Mammaglobin-1; Mammaglobin-A; MGB1; MGC71974; prostatein; prostatein C3 subunit; prostatein peptide C3; prostatic steroid binding protein, allele C3(1); prostatic steroid-binding protein C3; Psbpc3; SCGB2A2; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 2; UGB2 | |
Recombinant full-length protein | |
7 mL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
304-1A5 | |
tris with BSA, NP-40 and <0.1% sodium azide | |
Q13296 | |
Mouse | |
Protein A | |
RUO | |
4250 | |
2-8°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.