Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mammaglobin A Rabbit anti-Human, Clone: MGB/2123R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
Antigen | Mammaglobin A |
---|---|
Clone | MGB/2123R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
Mammaglobin A Monoclonal specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Mammaglobin A | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
Recombinant full-length human Mammaglobin A protein | |
Primary | |
Store at 4°C. |
MGB/2123R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
4250 | |
IgG κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title