Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mammaglobin A Recombinant Rabbit Monoclonal Antibody (PD00-74)
GREENER_CHOICE

Catalog No. FEMA550727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550727 Supplier Invitrogen™ Supplier No. MA550727
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Human skin tissue, human breast tissue. Predicted band size: 10 kDa Subcellular Location: Secreted.

Mammaglobin is a gene that is expressed almost exclusively in the normal breast epithelium and human breast cancer. It is a member of the secretoglobin gene family and forms a heterodimer with lipophilin B. It has been suggested that mammaglobin may be a useful marker for breast cancer clinical research. Studies investigating the detection of mRNA by RT PCR from circulating carcinoma cells in the peripheral blood of breast cancer patients have shown that mammaglobin is a highly specific marker and correlates with several prognostic factors, such as lymph node involvement.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mammaglobin A
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone PD00-74
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SCGB2A2
Gene Accession No. Q13296
Gene Alias C3.1; HGNC:7050; mammaglobin 1; mammaglobin A; Mammaglobin-1; Mammaglobin-A; MGB1; MGC71974; prostatein; prostatein C3 subunit; prostatein peptide C3; prostatic steroid binding protein, allele C3(1); prostatic steroid-binding protein C3; Psbpc3; SCGB2A2; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 2; UGB2
Gene Symbols SCGB2A2
Host Species Rabbit
Immunogen Synthetic peptide within C terminal Human Mammaglobin A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.