Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mammaglobin B Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297784JF549
Description
Mammaglobin B Polyclonal specifically detects Mammaglobin B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Mammaglobin B | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Lacryglobin, LIPHC, lipophilin C, lipophilin-C, LPHC, mammaglobin 2, mammaglobin B, Mammaglobin-2, MGB2mammaglobin-B, MGC71973, Secretoglobin family 2A member 1, secretoglobin, family 2A, member 1, UGB3mammaglobin-2 | |
Produced in rabbits immunized with E. coli-derived Human Mammaglobin B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
4246 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction