Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mammaglobin B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556981 Supplier Invitrogen™ Supplier No. PA556981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLLEDMVEKT INSDISIPEY KELLQEFIDS DAAAEAMGKF KQCFLNQSHR TLKNFGLMMH TVYDSIWCNM K Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 32%.

Secretoglobins (also called lipophilins or mammaglobins) are small secreted proteins of ndocrine-responsive organs and mucosal epithelia that form multimeric complexes and correlate with the development of various human cancers. Lipophilin A and Lipophilin B are orthologs of prostatein estramustine- binding protein), the major secretory glycoprotein of the rat ventral prostate gland. Lipophilin A, also designated LIPA, LPHA and secretoglobin, family 1D, member 1 (SCGB1D1), is a component of a heterodimeric molecule present in human tears. Lipophilin B, also designated LIPB, LPHB and secretoglobin, family 1D, member 2 (SCGB1D2), mRNA can be overexpressed in breast tumors and shows a high degree of correlation with the mRNA expression profile of mammaglobin. Histological detection in breast tissue of Mammaglobin A, also designated MGB1 and secretoglobin, family 2A, member 2 (SCGB2A2) and Mammaglobin B, also designated MGB2, Lipophilin C, LPHC, UGB3 and SCGB2A2, is a reliable diagnostic marker for breast tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mammaglobin B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCGB2A1
Gene Accession No. O75556
Gene Alias C3.1; Lacryglobin; LIPHC; lipophilin C; lipophilin-C; LPHC; LPNC; mammaglobin 2; mammaglobin B; mammaglobin-2; Mammaglobin-B; MGB2; prostatein; Prostatein peptide C3; Prostatic steroid-binding protein C3; Psbp1; Psbpc3; SCGB2A1; Scgb2a2; Secretoglobin family 2A member 1; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 1; UGB3
Gene Symbols SCGB2A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Mammaglobin B. Recombinant protein control fragment (Product #RP-95119).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4246
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.