Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mammaglobin B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599173
Description
Antibody detects endogenous levels of total Mammaglobin B.
Secretoglobins (also called lipophilins or mammaglobins) are small secreted proteins of ndocrine-responsive organs and mucosal epithelia that form multimeric complexes and correlate with the development of various human cancers. Lipophilin A and Lipophilin B are orthologs of prostatein estramustine- binding protein), the major secretory glycoprotein of the rat ventral prostate gland. Lipophilin A, also designated LIPA, LPHA and secretoglobin, family 1D, member 1 (SCGB1D1), is a component of a heterodimeric molecule present in human tears. Lipophilin B, also designated LIPB, LPHB and secretoglobin, family 1D, member 2 (SCGB1D2), mRNA can be overexpressed in breast tumors and shows a high degree of correlation with the mRNA expression profile of mammaglobin. Histological detection in breast tissue of Mammaglobin A, also designated MGB1 and secretoglobin, family 2A, member 2 (SCGB2A2) and Mammaglobin B, also designated MGB2, Lipophilin C, LPHC, UGB3 and SCGB2A2, is a reliable diagnostic marker for breast tumors.
Specifications
Mammaglobin B | |
Polyclonal | |
Unconjugated | |
SCGB2A1 | |
C3.1; Lacryglobin; LIPHC; lipophilin C; lipophilin-C; LPHC; LPNC; mammaglobin 2; mammaglobin B; mammaglobin-2; Mammaglobin-B; MGB2; prostatein; Prostatein peptide C3; Prostatic steroid-binding protein C3; Psbp1; Psbpc3; SCGB2A1; Scgb2a2; Secretoglobin family 2A member 1; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 1; UGB3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4246 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75556 | |
SCGB2A1 | |
A synthesized peptide derived from human SCGB2A1(Accession O75556), corresponding to amino acid residues L3-D53. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction