Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mammaglobin Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1919A1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1919A1 500 μL Concentrated
MS1919R1 1 mL Prediluted
MS1919RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1919A1 Supplier Epredia Supplier No. MS1919A1

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mammaglobin, Mouse Monoclonal Antibody.

Mammaglobin, a member of the uteroglobin family, is a 10kDa glycoprotein. Mammaglobin mRNA expression is breast tissue specific. It is overexpressed in human breast cancer. The use of Mammaglobin RT-PCR has been demonstrated to detect disseminated breast cancer. In a study to evaluate the frequency and specificity of mammaglobin, 80% of all primary and metastatic breast tumors tested were immunopositive irrespective of tumor grade, while mammaglobin mRNA was detected in 60% of the peripheral stem cell collections from breast cancer patients.

Host Species: Mouse

Clone: 304-1A5

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant full-length protein

Molecular Weight: 10kDa

Positive Control: Breast carcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Mammaglobin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 304-1A5
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant full-length protein
Quantity 500 μL
Regulatory Status ASR
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.