Learn More
Description
Host Species: Mouse
Clone: 304-1A5
Isotype: IgG1
Species Reactivity: Human. Others not tested.
Immunogen: Recombinant full-length protein
Molecular Weight: 10kDa
Positive Control: Breast carcinoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Mammaglobin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 304-1A5 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant full-length protein |
| Quantity | 1 mL |
| Regulatory Status | ASR |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.