Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAMSTR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565581
Description
Immunogen sequence: PPGPSLWEGTD SQQPHPRMKP SPLTPCPPGV PSPSPPPHKL ELQTLKLEEL TVSELRQ Highest antigen sequence identity to the following orthologs - mouse 74%, rat 74%.
Transcriptional coactivator. Stimulates the transcriptional activity of MEF2C. Stimulates MYOD1 activity in part via MEF2, resulting in an enhancement of skeletal muscle differentiation.
Specifications
MAMSTR | |
Polyclonal | |
Unconjugated | |
MAMSTR | |
likely ortholog of MEF2-activating SAP transcriptional regulator; MAMSTR; MASTR; MEF2 activating motif and SAP domain containing transcriptional regulator; MEF2-activating motif and SAP domain-containing transcriptional regulator; MEF2-activating SAP transcriptional regulatory protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284358 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZN01 | |
MAMSTR | |
Recombinant Human MAMSTR. Recombinant protein control fragment (Product #RP-99168). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction