Learn More
Invitrogen™ MAN1 Monoclonal Antibody (OTI3H1)

Description
MAN1 Monoclonal Antibody for Western Blot, ICC/IF, Flow, IP
Specifications
Specifications
| Antigen | MAN1 |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI3H1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LEMD3 |
| Gene Accession No. | Q9Y2U8 |
| Gene Alias | AI316861; inner nuclear membrane protein Man1; integral inner nuclear membrane protein; integral inner nuclear membrane protein MAN1; LEM domain containing 3; LEM domain-containing protein 3; LEMD3; MAN1; RGD1561001 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.