Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572734
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572734 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572734 Supplier Invitrogen™ Supplier No. PA572734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAN1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Can function as a specific repressor of TGF-beta, activin, and BMP signaling through its interaction with the R-SMAD proteins. Antagonizes TGF-beta-induced cell proliferation arrest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LEMD3
Gene Accession No. Q9WU40, Q9Y2U8
Gene Alias AI316861; inner nuclear membrane protein Man1; integral inner nuclear membrane protein; integral inner nuclear membrane protein MAN1; LEM domain containing 3; LEM domain-containing protein 3; LEMD3; MAN1; RGD1561001
Gene Symbols LEMD3
Host Species Rabbit
Immunogen LEMD3 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human LEMD3. The immunogen is within the last 50 amino acids of LEMD3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23592, 380664
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.