Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555227 Supplier Invitrogen™ Supplier No. PA555227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGQAFHLDRR NSPPNSLTPC LKIRNMFDPV MEIGDQWHLA IQEAILEKCS DNDGIVHIAV DKNSREGCVY VKCLSPEYAG KAFKALHGSW FDGKLVTVKY LRLDRYHHRF PQALTSNTPL KPSNKHMNSM SHLRLRT Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

Can function as a specific repressor of TGF-beta, activin, and BMP signaling through its interaction with the R-SMAD proteins. Antagonizes TGF-beta-induced cell proliferation arrest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LEMD3
Gene Accession No. Q9Y2U8
Gene Alias AI316861; inner nuclear membrane protein Man1; integral inner nuclear membrane protein; integral inner nuclear membrane protein MAN1; LEM domain containing 3; LEM domain-containing protein 3; LEMD3; MAN1; RGD1561001
Gene Symbols LEMD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LEMD3. Recombinant protein control fragment (Product #RP-102454).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23592
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.