Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAN1A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564122
Description
Immunogen sequence: MKHEFEEAKS WVEENLDFNV NAEISVFEV Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
This gene encodes a class I mammalian Golgi 1,2-mannosidase which is a type II transmembrane protein. This protein catalyzes the hydrolysis of three terminal mannose residues from peptide-bound Man(9)-GlcNAc(2) oligosaccharides and belongs to family 47 of glycosyl hydrolases.
Specifications
MAN1A1 | |
Polyclonal | |
Unconjugated | |
MAN1A1 | |
Aa2-166; Alpha-1,2-mannosidase IA; HUMM3; HUMM9; Man(9)-alpha-mannosidase; Man1a; MAN1A1; MAN9; Man9-mannosidase; mannosidase 1, alpha; mannosidase alpha class 1A member 1; mannosidase, alpha, class 1A, member 1; mannosyl-oligosaccharide 1,2-alpha-mannosidase 1A; mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; LOW QUALITY PROTEIN: mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; mannosyl-oligosaccharide 1,2-alpha-mannosidase IA-like; mannosyl-oligosaccharide alpha-1,2-mannosidase; PCR1; processing alpha-1,2-mannosidase IA; si:dkey-164k19.1; si:dkeyp-6c7.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4121 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P33908 | |
MAN1A1 | |
Recombinant protein corresponding to Human MAN1A1. Recombinant protein control fragment (Product #RP-103516). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction